Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144966.5 | NP_659403.4 | 12 - 13 | 2901 | 123 |
XM_005251382.3 | XP_005251439.1 | 11 - 12 | 3149 | 123 | |
XM_017014316.1 | XP_016869805.1 | 11 - 12 | 3176 | 123 | |
XM_017014317.1 | XP_016869806.1 | 12 - 13 | 2719 | 123 | |
XM_017014318.1 | XP_016869807.1 | 12 - 13 | 3474 | 123 | |
XM_017014319.1 | XP_016869808.1 | 13 - 14 | 3378 | 123 | |
XM_017014320.1 | XP_016869809.1 | 12 - 13 | 3305 | 123 | |
XM_017014321.1 | XP_016869810.1 | 11 - 12 | 2968 | 123 | |
XM_017014322.1 | XP_016869811.1 | 11 - 12 | 2762 | 123 | |
XM_017014323.1 | XP_016869812.1 | 11 - 12 | 2435 | 123 | |
XM_017014324.1 | XP_016869813.1 | 11 - 12 | 3176 | 123 | |
XM_017014325.1 | XP_016869814.1 | 11 - 12 | 3176 | 123 | |
XM_017014326.1 | XP_016869815.1 | 10 - 11 | 1992 | 123 | |
XM_017014327.1 | XP_016869816.1 | 7 - 8 | 1361 | 123 | |
XM_017014328.1 | XP_016869817.1 | 11 - 12 | 3176 | 123 | |
XM_017014329.1 | XP_016869818.1 | 11 - 12 | 3176 | 123 | |
XM_017014330.1 | XP_016869819.1 | 11 - 12 | 3176 | 123 | |
GenBank mRNA | AB160987.1 | - | 10 - 11 | 123 | |
BX537574.1 | - | 12 - 13 | 123 |