Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144966.5 | NP_659403.4 | 23 - 24 | 4794 | 124 |
XM_005251382.3 | XP_005251439.1 | 22 - 23 | 5042 | 124 | |
XM_006716729.3 | XP_006716792.1 | 2 - 3 | 283 | 124 | |
XM_017014316.1 | XP_016869805.1 | 22 - 23 | 5069 | 124 | |
XM_017014317.1 | XP_016869806.1 | 23 - 24 | 4612 | 124 | |
XM_017014318.1 | XP_016869807.1 | 23 - 24 | 5367 | 124 | |
XM_017014319.1 | XP_016869808.1 | 24 - 25 | 5271 | 124 | |
XM_017014320.1 | XP_016869809.1 | 23 - 24 | 5198 | 124 | |
XM_017014321.1 | XP_016869810.1 | 22 - 23 | 4861 | 124 | |
XM_017014322.1 | XP_016869811.1 | 22 - 23 | 4655 | 124 | |
XM_017014323.1 | XP_016869812.1 | 22 - 23 | 4328 | 124 | |
XM_017014324.1 | XP_016869813.1 | 22 - 23 | 5069 | 124 | |
XM_017014325.1 | XP_016869814.1 | 22 - 23 | 5069 | 124 | |
XM_017014326.1 | XP_016869815.1 | 21 - 22 | 3885 | 124 | |
XM_017014327.1 | XP_016869816.1 | 18 - 19 | 3254 | 124 | |
XM_017014328.1 | XP_016869817.1 | 22 - 23 | 5069 | 124 | |
XM_017014329.1 | XP_016869818.1 | 22 - 23 | 5069 | 124 | |
GenBank mRNA | AB160987.1 | - | 21 - 22 | 3978 | 124 |