Assay Design



Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011517281.1 XP_011515583.1 1 - 2 91 78
XM_011517282.1 XP_011515584.1 1 - 2 91 78

View Full Product Details