Assay Design



Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011517271.2 XP_011515573.1 42 - 43 5280 108
XM_017013802.1 XP_016869291.1 41 - 42 5252 108
GenBank mRNA AL834145.1 - 25 - 26 3401 108

View Full Product Details