Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006768.4 | NP_006759.3 | 2 - 3 | 451 | 95 |
XM_005253944.4 | XP_005254001.1 | 2 - 3 | 551 | 95 | |
XM_011538789.2 | XP_011537091.1 | 2 - 3 | 370 | 95 | |
GenBank mRNA | AB208878.1 | - | 2 - 3 | 406 | 95 |
AF035620.1 | - | 2 - 3 | 386 | 95 | |
AK290379.1 | - | 2 - 3 | 391 | 95 | |
AK308279.1 | - | 2 - 3 | 298 | 95 | |
AY332222.1 | - | 2 - 3 | 386 | 95 | |
BC136698.1 | - | 2 - 3 | 386 | 95 | |
BC136699.1 | - | 2 - 3 | 386 | 95 |