Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080874.3 | NP_543150.1 | 1 - 2 | 312 | 80 |
XM_005262759.1 | XP_005262816.1 | 3 - 4 | 741 | 80 | |
GenBank mRNA | AY057053.1 | - | 1 - 2 | 198 | 80 |
BC065710.1 | - | 1 - 2 | 219 | 80 | |
BX647857.1 | - | 1 - 2 | 313 | 80 |