Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039091.2 | NP_001034180.1 | 4 - 5 | 689 | 91 |
NM_002765.4 | NP_002756.1 | 4 - 5 | 680 | 91 | |
GenBank mRNA | AK313246.1 | - | 4 - 5 | 652 | 91 |
BC030019.2 | - | 4 - 5 | 621 | 91 | |
BC040483.1 | - | 4 - 5 | 623 | 91 | |
BC110875.1 | - | 4 - 5 | 614 | 91 | |
BC119662.1 | - | 4 - 5 | 621 | 91 | |
BC119663.1 | - | 4 - 5 | 612 | 91 | |
CR988262.1 | - | 4 - 5 | 638 | 91 | |
Y00971.1 | - | 4 - 5 | 597 | 91 |