Assay Design
Gene Transcripts
- Gene Symbol
- MYH11
- Entrez Gene ID
-
4629
- Gene Name
- myosin heavy chain 11
- Gene Aliases
-
AAT4, FAA4, SMHC, SMMHC
- Chromosome Location
-
Chr.16: 15703135 - 15857032 on Build GRCh38
- Gene Symbol
- MYH9
- Entrez Gene ID
-
4627
- Gene Name
- myosin, heavy chain 9, non-muscle
- Gene Aliases
-
BDPLT6, DFNA17, EPSTS, FTNS, MHA, NMHC-II-A, NMMHC-IIA, NMMHCA
- Chromosome Location
-
Chr.22: 36281277 - 36388067 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KU159427.1 |
- |
6 - 7 |
1770 |
103 |
View Full Product Details