Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040113.1 | NP_001035202.1 | 40 - 41 | 5741 | 57 |
NM_001040114.1 | NP_001035203.1 | 40 - 41 | 5741 | 57 | |
NM_002474.2 | NP_002465.1 | 39 - 40 | 5720 | 57 | |
NM_022844.2 | NP_074035.1 | 39 - 40 | 5720 | 57 | |
XM_011522502.2 | XP_011520804.1 | 39 - 40 | 5722 | 57 | |
XM_017023250.1 | XP_016878739.1 | 40 - 41 | 5705 | 57 | |
GenBank mRNA | AB020673.2 | - | 39 - 40 | 5688 | 57 |
AF013570.1 | - | 10 - 11 | 1564 | 57 | |
AF020091.1 | - | 10 - 11 | 1564 | 57 | |
AK311080.1 | - | 11 - 12 | 1901 | 57 | |
AY520816.1 | - | 40 - 41 | 5722 | 57 | |
AY520817.1 | - | 40 - 41 | 5722 | 57 | |
BC080545.1 | - | 8 - 9 | 1193 | 57 | |
BC101677.1 | - | 39 - 40 | 5684 | 57 | |
BC104906.1 | - | 39 - 40 | 5684 | 57 | |
BC143364.1 | - | 40 - 41 | 5705 | 57 | |
BX641087.1 | - | 10 - 11 | 1184 | 57 | |
CR749516.1 | - | 6 - 7 | 3533 | 57 | |
D10667.1 | - | 18 - 19 | 2955 | 57 | |
EU489063.1 | - | 40 - 41 | 5741 | 57 | |
X69292.1 | - | 15 - 16 | 2341 | 57 |