Assay Design



Gene Transcripts

Gene Symbol
STMN2
Entrez Gene ID
11075
Gene Name
stathmin 2
Gene Aliases
SCG10, SCGN10
Chromosome Location
Chr.8: 79610814 - 79666175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199214.1 NP_001186143.1 3 - 4 673 69
NM_007029.3 NP_008960.2 3 - 4 673 69
XM_005251142.2 XP_005251199.1 4 - 5 297 69
GenBank mRNA AK292737.1 - 3 - 4 345 69
AK297594.1 - 4 - 5 334 69
AK309130.1 - 3 - 4 560 69
BC006302.2 - 3 - 4 342 69
BQ069488.1 - 3 - 4 335 69
BT020034.1 - 2 - 3 291 69
CR456833.1 - 2 - 3 291 69
D50375.1 - 3 - 4 408 69

View Full Product Details