Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053002.5 | NP_443728.3 | 35 - 36 | 5470 | 63 |
XM_006713487.3 | XP_006713550.1 | 37 - 38 | 5847 | 63 | |
XM_011512390.2 | XP_011510692.1 | 36 - 37 | 5742 | 63 | |
XM_011512394.2 | XP_011510696.1 | 37 - 38 | 5847 | 63 | |
XM_017005676.1 | XP_016861165.1 | 37 - 38 | 5847 | 63 | |
XM_017005677.1 | XP_016861166.1 | 37 - 38 | 5844 | 63 | |
XM_017005678.1 | XP_016861167.1 | 37 - 38 | 5847 | 63 | |
XM_017005679.1 | XP_016861168.1 | 36 - 37 | 5577 | 63 | |
XM_017005680.1 | XP_016861169.1 | 35 - 36 | 5565 | 63 | |
XM_017005681.1 | XP_016861170.1 | 22 - 23 | 3233 | 63 | |
GenBank mRNA | AB046855.1 | - | 22 - 23 | 3213 | 63 |
AF399708.1 | - | 35 - 36 | 5379 | 63 | |
BC144559.1 | - | 33 - 34 | 4954 | 63 |