Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053002.5 | NP_443728.3 | 23 - 24 | 3579 | 77 |
XM_006713487.3 | XP_006713550.1 | 25 - 26 | 3956 | 77 | |
XM_011512390.2 | XP_011510692.1 | 24 - 25 | 3851 | 77 | |
XM_011512394.2 | XP_011510696.1 | 25 - 26 | 3956 | 77 | |
XM_011512399.2 | XP_011510701.1 | 25 - 26 | 3956 | 77 | |
XM_017005676.1 | XP_016861165.1 | 25 - 26 | 3956 | 77 | |
XM_017005677.1 | XP_016861166.1 | 25 - 26 | 3953 | 77 | |
XM_017005678.1 | XP_016861167.1 | 25 - 26 | 3956 | 77 | |
XM_017005679.1 | XP_016861168.1 | 24 - 25 | 3686 | 77 | |
XM_017005680.1 | XP_016861169.1 | 23 - 24 | 3674 | 77 | |
XM_017005681.1 | XP_016861170.1 | 10 - 11 | 1342 | 77 | |
GenBank mRNA | AB046855.1 | - | 10 - 11 | 1322 | 77 |
AF399708.1 | - | 23 - 24 | 3488 | 77 | |
AK022714.1 | - | 7 - 8 | 1028 | 77 | |
BC144559.1 | - | 21 - 22 | 3063 | 77 |