Assay Design



Gene Transcripts

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
Chromosome Location
Chr.1: 91949371 - 92014428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242805.2 NP_001229734.2 19 - 20 3192 97
NM_001242806.2 NP_001229735.2 18 - 19 3135 97
NM_001242807.2 NP_001229736.2 17 - 18 2791 97
NM_001242808.2 NP_001229737.2 17 - 18 2985 97
NM_001242810.2 NP_001229739.2 18 - 19 3479 97
NM_001726.4 NP_001717.3 18 - 19 2918 97
NM_207189.3 NP_997072.2 18 - 19 3123 97
XM_006710853.3 XP_006710916.1 19 - 20 3708 97
XM_006710854.3 XP_006710917.1 20 - 21 3553 97
XM_006710855.3 XP_006710918.1 20 - 21 3558 97
XM_006710856.3 XP_006710919.1 20 - 21 3532 97
XM_006710857.3 XP_006710920.1 20 - 21 3522 97
XM_011542032.2 XP_011540334.1 20 - 21 3042 97
XM_011542033.2 XP_011540335.1 20 - 21 3016 97
XM_011542034.2 XP_011540336.1 20 - 21 3032 97
XM_011542035.2 XP_011540337.1 20 - 21 3006 97
XM_011542036.2 XP_011540338.1 18 - 19 2921 97
GenBank mRNA AK302758.1 - 17 - 18 2727 97
AK303008.1 - 18 - 19 2883 97
AK316442.1 - 17 - 18 2790 97
AY338951.1 - 19 - 20 3192 97
BX648135.1 - 18 - 19 3214 97

View Full Product Details