Assay Design



Gene Transcripts

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
Chromosome Location
Chr.1: 91949371 - 92014428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242805.2 NP_001229734.2 4 - 5 749 105
NM_001242806.2 NP_001229735.2 3 - 4 680 105
NM_001242810.2 NP_001229739.2 3 - 4 1036 105
NM_001726.4 NP_001717.3 3 - 4 475 105
NM_207189.3 NP_997072.2 3 - 4 680 105
XM_006710853.3 XP_006710916.1 4 - 5 1265 105
XM_006710854.3 XP_006710917.1 5 - 6 1110 105
XM_006710855.3 XP_006710918.1 5 - 6 1115 105
XM_006710856.3 XP_006710919.1 5 - 6 1089 105
XM_006710857.3 XP_006710920.1 5 - 6 1079 105
XM_011542032.2 XP_011540334.1 5 - 6 599 105
XM_011542033.2 XP_011540335.1 5 - 6 573 105
XM_011542034.2 XP_011540336.1 5 - 6 589 105
XM_011542035.2 XP_011540337.1 5 - 6 563 105
XM_011542036.2 XP_011540338.1 3 - 4 478 105
GenBank mRNA AF019085.1 - 3 - 4 438 105
AY338951.1 - 4 - 5 749 105
BC005281.1 - 3 - 4 406 105
BC017582.1 - 3 - 4 481 105
BC047900.1 - 3 - 4 445 105
BC062700.1 - 3 - 4 440 105
BG717291.1 - 3 - 4 471 105
BX648135.1 - 3 - 4 771 105
DB072781.1 - 3 - 4 431 105
DC401352.1 - 3 - 4 476 105
DC402057.1 - 3 - 4 485 105

View Full Product Details