Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242805.2 | NP_001229734.2 | 7 - 8 | 1381 | 137 |
NM_001242806.2 | NP_001229735.2 | 6 - 7 | 1324 | 137 | |
NM_001242807.2 | NP_001229736.2 | 5 - 6 | 980 | 137 | |
NM_001242808.2 | NP_001229737.2 | 5 - 6 | 1174 | 137 | |
NM_001242810.2 | NP_001229739.2 | 6 - 7 | 1668 | 137 | |
NM_001726.4 | NP_001717.3 | 6 - 7 | 1107 | 137 | |
NM_207189.3 | NP_997072.2 | 6 - 7 | 1312 | 137 | |
XM_006710853.3 | XP_006710916.1 | 7 - 8 | 1897 | 137 | |
XM_006710854.3 | XP_006710917.1 | 8 - 9 | 1742 | 137 | |
XM_006710855.3 | XP_006710918.1 | 8 - 9 | 1747 | 137 | |
XM_006710856.3 | XP_006710919.1 | 8 - 9 | 1721 | 137 | |
XM_006710857.3 | XP_006710920.1 | 8 - 9 | 1711 | 137 | |
XM_011542032.2 | XP_011540334.1 | 8 - 9 | 1231 | 137 | |
XM_011542033.2 | XP_011540335.1 | 8 - 9 | 1205 | 137 | |
XM_011542034.2 | XP_011540336.1 | 8 - 9 | 1221 | 137 | |
XM_011542035.2 | XP_011540337.1 | 8 - 9 | 1195 | 137 | |
XM_011542036.2 | XP_011540338.1 | 6 - 7 | 1110 | 137 | |
GenBank mRNA | AF019085.1 | - | 6 - 7 | 1070 | 137 |
AK302758.1 | - | 5 - 6 | 916 | 137 | |
AK303008.1 | - | 6 - 7 | 1072 | 137 | |
AK316442.1 | - | 5 - 6 | 979 | 137 | |
AY338951.1 | - | 7 - 8 | 1381 | 137 | |
BC005281.1 | - | 6 - 7 | 1038 | 137 | |
BC017582.1 | - | 6 - 7 | 1113 | 137 | |
BC047900.1 | - | 6 - 7 | 1077 | 137 | |
BC062700.1 | - | 6 - 7 | 1072 | 137 | |
BX648135.1 | - | 6 - 7 | 1403 | 137 |