Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005245093.3 | XP_005245150.1 | 2 - 3 | 284 | 118 |
XM_017001052.1 | XP_016856541.1 | 1 - 2 | 414 | 118 | |
XM_017001053.1 | XP_016856542.1 | 1 - 2 | 414 | 118 | |
XM_017001054.1 | XP_016856543.1 | 2 - 3 | 284 | 118 | |
GenBank mRNA | BC005253.1 | - | 1 - 2 | 86 | 118 |