Assay Design



Gene Transcripts

Gene Symbol
GTF2B
Entrez Gene ID
2959
Gene Name
general transcription factor IIB
Gene Aliases
TF2B, TFIIB
Chromosome Location
Chr.1: 88852638 - 88891944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011541299.1 XP_011539601.1 1 - 2 344 107

View Full Product Details