Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005459.3 | NP_005450.3 | 1 - 2 | 110 | |
XM_005247910.3 | XP_005247967.1 | 1 - 2 | 110 | ||
GenBank mRNA | AB593147.1 | - | 1 - 2 | 221 | 110 |
AF110002.1 | - | 1 - 2 | 217 | 110 | |
AF110003.1 | - | 1 - 2 | 110 | ||
AV732101.1 | - | 1 - 2 | 110 | ||
BC103993.1 | - | 1 - 2 | 217 | 110 | |
BC103994.1 | - | 1 - 2 | 217 | 110 |