Assay Design
Gene Transcripts
- Gene Symbol
- CCDC39
- Entrez Gene ID
-
339829
- Gene Name
- coiled-coil domain containing 39
- Gene Aliases
-
CILD14, FAP59
- Chromosome Location
-
Chr.3: 180614008 - 180679495 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_181426.1 |
NP_852091.1 |
8 - 9 |
1148 |
152 |
GenBank mRNA |
AL122120.1 |
- |
14 - 15 |
1905 |
152 |
View Full Product Details