Assay Design



Gene Transcripts

Gene Symbol
CCDC39
Entrez Gene ID
339829
Gene Name
coiled-coil domain containing 39
Gene Aliases
CILD14, FAP59
Chromosome Location
Chr.3: 180614008 - 180679495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181426.1 NP_852091.1 7 - 8 1042 83
GenBank mRNA AK304270.1 - 7 - 8 1021 83
AL122120.1 - 13 - 14 1799 83

View Full Product Details