Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318050.1 | NP_001304979.1 | 5 - 6 | 1430 | 157 |
NM_138969.3 | NP_620419.2 | 6 - 7 | 1562 | 157 | |
GenBank mRNA | AB083038.1 | - | 6 - 7 | 1468 | 157 |
AK057667.1 | - | 6 - 7 | 905 | 157 | |
AK294634.1 | - | 5 - 6 | 833 | 157 | |
AY444559.1 | - | 5 - 6 | 829 | 157 | |
BC037219.1 | - | 6 - 7 | 909 | 157 | |
BC064525.1 | - | 6 - 7 | 881 | 157 |