Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020761.2 | NP_065812.1 | 14 - 15 | 2395 | 91 |
GenBank mRNA | AB037724.1 | - | 10 - 11 | 1077 | 91 |
AB082951.1 | - | 14 - 15 | 1590 | 91 | |
AK055912.1 | - | 14 - 15 | 2241 | 91 | |
AK296321.1 | - | 14 - 15 | 2390 | 91 | |
AY090663.1 | - | 14 - 15 | 1952 | 91 | |
BC136652.1 | - | 14 - 15 | 1785 | 91 | |
BC136654.1 | - | 14 - 15 | 1785 | 91 |