Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163034.1 | NP_001156506.1 | 24 - 25 | 3700 | 93 |
NM_020761.2 | NP_065812.1 | 28 - 29 | 4174 | 93 | |
GenBank mRNA | AB037724.1 | - | 24 - 25 | 2856 | 93 |
AB082951.1 | - | 28 - 29 | 3369 | 93 | |
AY090663.1 | - | 28 - 29 | 3731 | 93 | |
BC033258.1 | - | 4 - 5 | 389 | 93 | |
BC136652.1 | - | 28 - 29 | 3564 | 93 | |
BC136654.1 | - | 28 - 29 | 3564 | 93 |