Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163034.1 | NP_001156506.1 | 3 - 4 | 1155 | 90 |
NM_020761.2 | NP_065812.1 | 3 - 4 | 1155 | 90 | |
GenBank mRNA | AB082951.1 | - | 3 - 4 | 350 | 90 |
AK055912.1 | - | 3 - 4 | 1001 | 90 | |
AK296321.1 | - | 3 - 4 | 1150 | 90 | |
AY090663.1 | - | 3 - 4 | 712 | 90 | |
BC064515.1 | - | 3 - 4 | 1103 | 90 | |
BC136652.1 | - | 3 - 4 | 545 | 90 | |
BC136654.1 | - | 3 - 4 | 545 | 90 |