Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160423.1 | NP_001153895.1 | 1 - 2 | 510 | 86 |
NM_006546.3 | NP_006537.3 | 1 - 2 | 510 | 86 | |
XM_011524201.2 | XP_011522503.1 | 1 - 2 | 461 | 86 | |
XM_017024022.1 | XP_016879511.1 | 1 - 2 | 461 | 86 | |
GenBank mRNA | AF117106.1 | - | 1 - 2 | 185 | 86 |
AF198254.1 | - | 1 - 2 | 481 | 86 | |
AK313229.1 | - | 1 - 2 | 481 | 86 | |
BP238821.1 | - | 1 - 2 | 510 | 86 | |
DA758608.1 | - | 1 - 2 | 510 | 86 | |
DQ227344.1 | - | 1 - 2 | 176 | 86 |