Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278942.1 | NP_001265871.1 | 1 - 2 | 175 | 61 |
NM_006753.5 | NP_006744.2 | 1 - 2 | 175 | 61 | |
NR_103874.1 | 1 - 2 | 175 | 61 | ||
GenBank mRNA | BC003001.1 | - | 1 - 2 | 102 | 61 |
BC006197.2 | - | 1 - 2 | 102 | 61 | |
BC014878.1 | - | 1 - 2 | 137 | 61 | |
BI820270.1 | - | 1 - 2 | 118 | 61 | |
BM451240.1 | - | 1 - 2 | 174 | 61 | |
CD104374.1 | - | 1 - 2 | 177 | 61 |