Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000031.5 | NP_000022.3 | 6 - 7 | 685 | 151 |
NM_001003945.2 | NP_001003945.1 | 6 - 7 | 851 | 151 | |
NM_001317745.1 | NP_001304674.1 | 5 - 6 | 694 | 151 | |
XM_011518364.2 | XP_011516666.1 | 7 - 8 | 877 | 151 | |
GenBank mRNA | AK131490.1 | - | 6 - 7 | 824 | 151 |
AK290490.1 | - | 6 - 7 | 797 | 151 | |
AK295945.1 | - | 5 - 6 | 694 | 151 | |
AK312552.1 | - | 6 - 7 | 633 | 151 | |
BC000977.3 | - | 6 - 7 | 609 | 151 | |
M13928.1 | - | 5 - 6 | 549 | 151 | |
S99468.1 | - | 5 - 6 | 529 | 151 | |
S99471.1 | - | 5 - 6 | 529 | 151 |