Assay Design



Gene Transcripts

Gene Symbol
NFASC
Entrez Gene ID
23114
Gene Name
neurofascin
Gene Aliases
NF, NRCAML
Chromosome Location
Chr.1: 204828652 - 205022822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005388.2 NP_001005388.2 1 - 2 130 153
NM_001005389.1 NP_001005389.2 1 - 2 130 153
NM_001160332.1 NP_001153804.1 1 - 2 130 153
NM_001160333.1 NP_001153805.1 1 - 2 130 153
NM_015090.3 NP_055905.2 1 - 2 130 153
XM_011509318.2 XP_011507620.1 1 - 2 132 153
XM_011509320.2 XP_011507622.1 1 - 2 132 153
XM_017000738.1 XP_016856227.1 1 - 2 132 153
XM_017000739.1 XP_016856228.1 1 - 2 132 153
XM_017000740.1 XP_016856229.1 1 - 2 132 153
XM_017000743.1 XP_016856232.1 1 - 2 132 153
GenBank mRNA BC040674.1 - 1 - 2 135 153
BG702675.1 - 1 - 2 135 153

View Full Product Details