Assay Design



Gene Transcripts

Gene Symbol
NFASC
Entrez Gene ID
23114
Gene Name
neurofascin
Gene Aliases
NF, NRCAML
Chromosome Location
Chr.1: 204828652 - 205022822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160332.1 NP_001153804.1 16 - 17 2194 144
NM_015090.3 NP_055905.2 16 - 17 2194 144
XM_005244991.3 XP_005245048.2 16 - 17 2274 144
XM_005244992.4 XP_005245049.2 16 - 17 2274 144
XM_005244993.3 XP_005245050.2 15 - 16 2223 144
XM_011509313.1 XP_011507615.1 15 - 16 1974 144
XM_011509315.1 XP_011507617.1 14 - 15 1923 144
XM_017000733.1 XP_016856222.1 16 - 17 2274 144
XM_017000734.1 XP_016856223.1 15 - 16 2223 144
XM_017000736.1 XP_016856225.1 15 - 16 1974 144
XM_017000738.1 XP_016856227.1 16 - 17 2196 144
XM_017000739.1 XP_016856228.1 16 - 17 2196 144
XM_017000740.1 XP_016856229.1 16 - 17 2196 144
GenBank mRNA AB018299.3 - 15 - 16 2025 144
AB177861.1 - 9 - 10 1259 144
BC137013.1 - 14 - 15 1931 144

View Full Product Details