Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003602.3 | 4 - 5 | 507 | 70 | |
GenBank mRNA | BC022013.2 | - | 4 - 5 | 461 | 70 |
BC067783.1 | - | 4 - 5 | 480 | 70 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_104007.1 | 3 - 4 | 369 | 70 | |
GenBank mRNA | BC070189.1 | - | 3 - 4 | 369 | 70 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif containing 74 | TRIM74 | TRIM50C | -1.8kb | Anti-sense | Chr. - 72954845 - 73005932 on Build GRCh38 |
Downstream | stromal antigen 3-like 3 (pseudogene) | STAG3L3 | STAG3L1, STAG3L2, STAG3L3P | 24.4kb | Anti-sense | Chr. - 72997295 - 73005943 on Build GRCh38 |