Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284277.1 | NP_001271206.1 | 9 - 10 | 765 | 73 |
NM_001284278.1 | NP_001271207.1 | 9 - 10 | 743 | 73 | |
NM_012399.4 | NP_036531.1 | 9 - 10 | 765 | 73 | |
XM_011530052.2 | XP_011528354.1 | 9 - 10 | 778 | 73 | |
GenBank mRNA | AK123384.1 | - | 2 - 3 | 199 | 73 |
AK302367.1 | - | 9 - 10 | 729 | 73 | |
AK313064.1 | - | 9 - 10 | 715 | 73 | |
BC018704.1 | - | 9 - 10 | 689 | 73 | |
BC031427.1 | - | 9 - 10 | 682 | 73 | |
BU147254.1 | - | 9 - 10 | 726 | 73 | |
CR456541.1 | - | 9 - 10 | 717 | 73 | |
D30037.1 | - | 9 - 10 | 696 | 73 |