Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163.3 | NP_001154.2 | 9 - 10 | 2246 | 92 |
XM_005251968.3 | XP_005252025.1 | 8 - 9 | 2244 | 92 | |
XM_011518617.2 | XP_011516919.1 | 9 - 10 | 2201 | 92 | |
XM_017014670.1 | XP_016870159.1 | 9 - 10 | 2490 | 92 | |
GenBank mRNA | AF029106.1 | - | 9 - 10 | 2168 | 92 |
AF047347.1 | - | 9 - 10 | 2209 | 92 | |
L04953.1 | - | 8 - 9 | 1583 | 92 |