Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018242.2 | NP_060712.2 | 4 - 5 | 543 | 87 |
GenBank mRNA | AK001709.1 | - | 4 - 5 | 543 | 87 |
AK222625.1 | - | 4 - 5 | 547 | 87 | |
AK225360.1 | - | 4 - 5 | 521 | 87 | |
AK301854.1 | - | 3 - 4 | 500 | 87 | |
AK302620.1 | - | 4 - 5 | 543 | 87 | |
AK310270.1 | - | 4 - 5 | 362 | 87 | |
BC010661.2 | - | 4 - 5 | 532 | 87 | |
BC050592.1 | - | 4 - 5 | 517 | 87 | |
BC058882.1 | - | 4 - 5 | 519 | 87 |