Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294340.1 | NP_001281269.1 | 5 - 6 | 1110 | 69 |
NM_144604.3 | NP_653205.3 | 4 - 5 | 1038 | 69 | |
XM_011522863.1 | XP_011521165.1 | 5 - 6 | 1110 | 69 | |
XM_017022934.1 | XP_016878423.1 | 5 - 6 | 1110 | 69 | |
GenBank mRNA | AK225807.1 | - | 4 - 5 | 1008 | 69 |
AK300254.1 | - | 5 - 6 | 1088 | 69 | |
AK302716.1 | - | 4 - 5 | 665 | 69 | |
BC001584.1 | - | 1 - 2 | 134 | 69 | |
BC050463.1 | - | 4 - 5 | 985 | 69 | |
DB007291.1 | - | 1 - 2 | 124 | 69 |