Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294340.1 | NP_001281269.1 | 6 - 7 | 1198 | 79 |
NM_144604.3 | NP_653205.3 | 5 - 6 | 1126 | 79 | |
XM_011522863.1 | XP_011521165.1 | 6 - 7 | 1198 | 79 | |
XM_017022934.1 | XP_016878423.1 | 6 - 7 | 1198 | 79 | |
GenBank mRNA | AK225807.1 | - | 5 - 6 | 1096 | 79 |
AK302716.1 | - | 5 - 6 | 753 | 79 | |
BC001584.1 | - | 2 - 3 | 222 | 79 | |
BC050463.1 | - | 5 - 6 | 1073 | 79 | |
DB007291.1 | - | 2 - 3 | 212 | 79 |