Assay Design



Gene Transcripts

Gene Symbol
ZC3H18
Entrez Gene ID
124245
Gene Name
zinc finger CCCH-type containing 18
Gene Aliases
NHN1
Chromosome Location
Chr.16: 88570381 - 88631964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001294340.1 NP_001281269.1 13 - 14 2316 105
NM_144604.3 NP_653205.3 12 - 13 2244 105
XM_011522863.1 XP_011521165.1 11 - 12 1929 105
XM_011522865.1 XP_011521167.1 5 - 6 686 105
GenBank mRNA AK091849.1 - 7 - 8 1071 105
AK122593.1 - 4 - 5 3162 106
AK225807.1 - 12 - 13 2214 105
AK300254.1 - 13 - 14 2294 105
BC001584.1 - 9 - 10 1340 105
BC050463.1 - 12 - 13 2191 105

View Full Product Details