Assay Design



Gene Transcripts

Gene Symbol
TRIM55
Entrez Gene ID
84675
Gene Name
tripartite motif containing 55
Gene Aliases
MURF-2, RNF29, muRF2
Chromosome Location
Chr.8: 66112667 - 66175485 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033058.2 NP_149047.2 5 - 6 1064 114
NM_184085.1 NP_908973.1 5 - 6 1064 114
NM_184086.1 NP_908974.1 5 - 6 1064 114
XM_005251316.4 XP_005251373.1 7 - 8 2089 114
XM_011517613.2 XP_011515915.1 7 - 8 2089 114
XM_011517614.2 XP_011515916.1 7 - 8 2012 114
XM_011517615.2 XP_011515917.1 7 - 8 2089 114
XM_011517616.2 XP_011515918.1 7 - 8 2089 114
XM_011517617.2 XP_011515919.1 5 - 6 1146 114
XM_017013908.1 XP_016869397.1 5 - 6 782 114
XM_017013909.1 XP_016869398.1 4 - 5 601 114
GenBank mRNA AJ243488.1 - 5 - 6 1064 114
AJ243489.1 - 5 - 6 1064 114
AJ291712.1 - 5 - 6 970 114
AJ431704.1 - 5 - 6 1064 114
AK091310.1 - 5 - 6 1214 114
AK091728.1 - 5 - 6 884 114
BC007750.2 - 5 - 6 970 114
BT007212.1 - 5 - 6 790 114

View Full Product Details