Assay Design



Gene Transcripts

Gene Symbol
APBA1
Entrez Gene ID
320
Gene Name
amyloid beta precursor protein binding family A member 1
Gene Aliases
D9S411E, LIN10, MINT1, X11, X11A, X11ALPHA
Chromosome Location
Chr.9: 69427532 - 69673012 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163.3 NP_001154.2 3 - 4 1576 83
XM_005251968.3 XP_005252025.1 3 - 4 1607 83
XM_011518617.2 XP_011516919.1 3 - 4 1531 83
XM_017014670.1 XP_016870159.1 3 - 4 1820 83
GenBank mRNA AF029106.1 - 3 - 4 1498 83
AF047347.1 - 3 - 4 1539 83
L04953.1 - 2 - 3 913 83

View Full Product Details