Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000042.2 | NP_000033.2 | 1 - 2 | 127 | 109 |
GenBank mRNA | AK298416.1 | - | 1 - 2 | 99 | 109 |
AK313838.1 | - | 1 - 2 | 83 | 109 | |
BC020703.1 | - | 1 - 2 | 85 | 109 | |
BC026283.1 | - | 1 - 2 | 99 | 109 | |
BG566874.1 | - | 1 - 2 | 127 | 109 | |
M62839.1 | - | 1 - 2 | 107 | 109 | |
S80305.1 | - | 1 - 2 | 77 | 109 | |
X53595.1 | - | 1 - 2 | 101 | 109 | |
X57847.1 | - | 1 - 2 | 101 | 109 | |
X58100.1 | - | 1 - 2 | 89 | 109 |