Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000042.2 | NP_000033.2 | 6 - 7 | 834 | 103 |
GenBank mRNA | AK298416.1 | - | 6 - 7 | 593 | 103 |
AK313838.1 | - | 6 - 7 | 790 | 103 | |
BC020703.1 | - | 6 - 7 | 792 | 103 | |
BC026283.1 | - | 6 - 7 | 806 | 103 | |
BM005736.1 | - | 1 - 2 | 189 | 103 | |
HM212425.1 | - | 5 - 6 | 718 | 103 | |
M62839.1 | - | 6 - 7 | 814 | 103 | |
S80305.1 | - | 6 - 7 | 784 | 103 | |
X53595.1 | - | 6 - 7 | 808 | 103 | |
X57847.1 | - | 6 - 7 | 808 | 103 | |
X58100.1 | - | 6 - 7 | 796 | 103 |