Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000042.2 | NP_000033.2 | 7 - 8 | 1042 | 97 |
GenBank mRNA | AK298416.1 | - | 7 - 8 | 801 | 97 |
AK313838.1 | - | 7 - 8 | 998 | 97 | |
BC020703.1 | - | 7 - 8 | 1000 | 97 | |
BC026283.1 | - | 7 - 8 | 1014 | 97 | |
BM005736.1 | - | 2 - 3 | 397 | 97 | |
HM212425.1 | - | 6 - 7 | 926 | 97 | |
M62839.1 | - | 7 - 8 | 1022 | 97 | |
S80305.1 | - | 7 - 8 | 992 | 97 | |
X53595.1 | - | 7 - 8 | 1016 | 97 | |
X57847.1 | - | 7 - 8 | 1016 | 97 | |
X58100.1 | - | 7 - 8 | 1004 | 97 |