Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080284.2 | NP_525023.2 | 4 - 5 | 630 | 165 |
XM_017024404.1 | XP_016879893.1 | 4 - 5 | 942 | 165 | |
XM_017024405.1 | XP_016879894.1 | 4 - 5 | 942 | 165 | |
GenBank mRNA | AF373290.1 | - | 3 - 4 | 458 | 165 |
AK308431.1 | - | 4 - 5 | 650 | 165 | |
AY028898.1 | - | 4 - 5 | 630 | 165 | |
BC125231.1 | - | 4 - 5 | 629 | 165 | |
BC125232.1 | - | 4 - 5 | 629 | 165 |