Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022968.1 | NP_075257.1 | 2 - 3 | 316 | 78 |
GenBank mRNA | AF073518.1 | - | 2 - 3 | 283 | 78 |
BC021174.1 | - | 2 - 3 | 315 | 78 | |
DB444407.1 | - | 2 - 3 | 78 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178087.1 | NP_001171558.1 | 2 - 3 | 310 | 78 |
GenBank mRNA | CD679521.1 | - | 2 - 3 | 190 | 78 |
DB454907.1 | - | 2 - 3 | 310 | 78 |