Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142617.1 | NP_001136089.1 | 10 - 11 | 1108 | 57 |
NM_001142618.1 | NP_001136090.1 | 10 - 11 | 1090 | 57 | |
NM_001142619.1 | NP_001136091.1 | 10 - 11 | 1081 | 57 | |
NM_001199040.1 | NP_001185969.1 | 10 - 11 | 1224 | 57 | |
NM_001199041.1 | NP_001185970.1 | 10 - 11 | 1235 | 57 | |
NM_001199042.1 | NP_001185971.1 | 10 - 11 | 1339 | 57 | |
NM_022369.3 | NP_071764.3 | 10 - 11 | 1044 | 57 | |
XM_011521883.1 | XP_011520185.1 | 10 - 11 | 1010 | 57 | |
XM_011521884.1 | XP_011520186.1 | 10 - 11 | 1151 | 57 | |
XM_011521885.2 | XP_011520187.1 | 10 - 11 | 985 | 57 | |
XM_017022478.1 | XP_016877967.1 | 10 - 11 | 959 | 57 | |
XM_017022479.1 | XP_016877968.1 | 10 - 11 | 1050 | 57 | |
XM_017022480.1 | XP_016877969.1 | 10 - 11 | 1074 | 57 | |
GenBank mRNA | AF352728.1 | - | 10 - 11 | 910 | 57 |
AF352729.1 | - | 10 - 11 | 1020 | 57 | |
AF370419.2 | - | 10 - 11 | 1016 | 57 | |
AK022603.1 | - | 6 - 7 | 594 | 57 | |
AK056125.1 | - | 10 - 11 | 1105 | 57 | |
AK091152.1 | - | 10 - 11 | 1134 | 57 | |
AK092227.1 | - | 5 - 6 | 571 | 57 | |
AK291966.1 | - | 10 - 11 | 980 | 57 | |
AK298921.1 | - | 10 - 11 | 979 | 57 | |
AK299191.1 | - | 10 - 11 | 1224 | 57 | |
AK309676.1 | - | 10 - 11 | 1319 | 57 | |
AY358748.1 | - | 10 - 11 | 1020 | 57 | |
AY359089.1 | - | 10 - 11 | 910 | 57 | |
BC025256.1 | - | 10 - 11 | 1016 | 57 | |
BX537413.1 | - | 10 - 11 | 986 | 57 |