Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142617.1 | NP_001136089.1 | 12 - 13 | 1336 | 78 |
NM_001142618.1 | NP_001136090.1 | 12 - 13 | 1318 | 78 | |
NM_001142619.1 | NP_001136091.1 | 12 - 13 | 1309 | 78 | |
NM_001199040.1 | NP_001185969.1 | 12 - 13 | 1452 | 78 | |
NM_001199041.1 | NP_001185970.1 | 12 - 13 | 1463 | 78 | |
NM_001199042.1 | NP_001185971.1 | 12 - 13 | 1567 | 78 | |
NM_022369.3 | NP_071764.3 | 12 - 13 | 1272 | 78 | |
XM_011521883.1 | XP_011520185.1 | 12 - 13 | 1238 | 78 | |
XM_011521884.1 | XP_011520186.1 | 12 - 13 | 1379 | 78 | |
XM_017022478.1 | XP_016877967.1 | 12 - 13 | 1187 | 78 | |
XM_017022479.1 | XP_016877968.1 | 12 - 13 | 1278 | 78 | |
XM_017022480.1 | XP_016877969.1 | 12 - 13 | 1302 | 78 | |
GenBank mRNA | AF352728.1 | - | 12 - 13 | 1138 | 78 |
AF352729.1 | - | 12 - 13 | 1248 | 78 | |
AF370419.2 | - | 12 - 13 | 1244 | 78 | |
AK022603.1 | - | 8 - 9 | 822 | 78 | |
AK092227.1 | - | 7 - 8 | 799 | 78 | |
AK291966.1 | - | 12 - 13 | 1208 | 78 | |
AK298921.1 | - | 12 - 13 | 1207 | 78 | |
AK298928.1 | - | 10 - 11 | 876 | 78 | |
AK299191.1 | - | 12 - 13 | 1452 | 78 | |
AK302932.1 | - | 12 - 13 | 1463 | 78 | |
AK309676.1 | - | 12 - 13 | 1547 | 78 | |
AK310878.1 | - | 11 - 12 | 1325 | 78 | |
AY358748.1 | - | 12 - 13 | 1248 | 78 | |
AY359089.1 | - | 12 - 13 | 1138 | 78 | |
BC025256.1 | - | 12 - 13 | 1244 | 78 | |
BX537413.1 | - | 12 - 13 | 1214 | 78 |