Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142617.1 | NP_001136089.1 | 18 - 19 | 2084 | 69 |
NM_001142618.1 | NP_001136090.1 | 18 - 19 | 2066 | 69 | |
NM_001142619.1 | NP_001136091.1 | 18 - 19 | 2057 | 69 | |
NM_001199040.1 | NP_001185969.1 | 18 - 19 | 2200 | 69 | |
NM_001199041.1 | NP_001185970.1 | 18 - 19 | 2211 | 69 | |
NM_001199042.1 | NP_001185971.1 | 18 - 19 | 2315 | 69 | |
NM_022369.3 | NP_071764.3 | 18 - 19 | 2020 | 69 | |
XM_011521883.1 | XP_011520185.1 | 18 - 19 | 1986 | 69 | |
XM_011521884.1 | XP_011520186.1 | 18 - 19 | 2127 | 69 | |
XM_017022478.1 | XP_016877967.1 | 18 - 19 | 1935 | 69 | |
XM_017022479.1 | XP_016877968.1 | 18 - 19 | 2026 | 69 | |
XM_017022480.1 | XP_016877969.1 | 18 - 19 | 2050 | 69 | |
GenBank mRNA | AF352728.1 | - | 18 - 19 | 1886 | 69 |
AF352729.1 | - | 18 - 19 | 1996 | 69 | |
AF370419.2 | - | 18 - 19 | 1992 | 69 | |
AK022603.1 | - | 14 - 15 | 1570 | 69 | |
AK056125.1 | - | 18 - 19 | 2081 | 69 | |
AK091152.1 | - | 18 - 19 | 2110 | 69 | |
AK092227.1 | - | 13 - 14 | 1547 | 69 | |
AK291966.1 | - | 18 - 19 | 1956 | 69 | |
AK298921.1 | - | 18 - 19 | 1955 | 69 | |
AK298928.1 | - | 16 - 17 | 1624 | 69 | |
AK299191.1 | - | 18 - 19 | 2200 | 69 | |
AK302932.1 | - | 18 - 19 | 2211 | 69 | |
AK309676.1 | - | 18 - 19 | 2295 | 69 | |
AY358748.1 | - | 18 - 19 | 1996 | 69 | |
AY359089.1 | - | 18 - 19 | 1886 | 69 | |
BC025256.1 | - | 18 - 19 | 1992 | 69 | |
BX537413.1 | - | 18 - 19 | 1962 | 69 |