Assay Design



Gene Transcripts

Gene Symbol
ZNRF3
Entrez Gene ID
84133
Gene Name
zinc and ring finger 3
Gene Aliases
BK747E2.3, RNF203
Chromosome Location
Chr.22: 28800657 - 29057488 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206998.1 NP_001193927.1 1 - 2 301 127
GenBank mRNA AB051436.1 - 1 - 2 166 127
CR456397.1 - 1 - 2 74 127

View Full Product Details