Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206998.1 | NP_001193927.1 | 9 - 9 | 6561 | 146 |
NM_032173.3 | NP_115549.2 | 9 - 9 | 6354 | 146 | |
XM_011530435.2 | XP_011528737.1 | 10 - 10 | 6550 | 146 | |
XM_011530436.2 | XP_011528738.1 | 11 - 11 | 6605 | 146 | |
XM_011530437.1 | XP_011528739.1 | 10 - 10 | 7275 | 146 | |
XM_011530438.2 | XP_011528740.1 | 10 - 10 | 6791 | 146 | |
XM_017028990.1 | XP_016884479.1 | 10 - 10 | 7046 | 146 | |
GenBank mRNA | AB051436.1 | - | 9 - 9 | 6427 | 146 |
AK022809.1 | - | 1 - 1 | 3177 | 146 | |
BC042614.1 | - | 1 - 1 | 3390 | 146 | |
BC051344.1 | - | 1 - 1 | 3482 | 146 |