Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014876.5 | NP_055691.1 | 2 - 3 | 776 | 71 |
XM_005261876.2 | XP_005261933.1 | 3 - 4 | 1083 | 71 | |
XM_005261877.2 | XP_005261934.1 | 3 - 4 | 979 | 71 | |
XM_005261878.2 | XP_005261935.1 | 3 - 4 | 1007 | 71 | |
XM_005261879.2 | XP_005261936.1 | 3 - 4 | 1102 | 71 | |
GenBank mRNA | AK291827.1 | - | 2 - 3 | 569 | 71 |
BC015026.2 | - | 2 - 3 | 610 | 71 | |
CR456473.1 | - | 2 - 3 | 455 | 71 | |
CR456923.1 | - | 2 - 3 | 309 | 71 | |
D31884.1 | - | 2 - 3 | 588 | 71 |