Assay Design



Gene Transcripts

Gene Symbol
GOLGB1
Entrez Gene ID
2804
Gene Name
golgin B1
Gene Aliases
GCP, GCP372, GOLIM1
Chromosome Location
Chr.3: 121663199 - 121749908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256486.1 NP_001243415.1 12 - 13 1734 81
NM_001256487.1 NP_001243416.1 12 - 13 1762 81
NM_001256488.1 NP_001243417.1 11 - 12 1639 81
NM_004487.4 NP_004478.3 12 - 13 1719 81
XM_005247371.4 XP_005247428.1 12 - 13 2194 81
XM_005247372.4 XP_005247429.1 12 - 13 1770 81
XM_005247373.1 XP_005247430.1 11 - 12 1586 81
XM_006713587.1 XP_006713650.1 12 - 13 1709 81
XM_006713588.1 XP_006713651.1 12 - 13 1694 81
XM_006713589.1 XP_006713652.1 12 - 13 1691 81
XM_006713590.1 XP_006713653.1 11 - 12 1586 81
XM_006713591.1 XP_006713654.1 10 - 11 1469 81
XM_011512699.2 XP_011511001.1 12 - 13 1661 81
XM_017006189.1 XP_016861678.1 12 - 13 2179 81
XM_017006190.1 XP_016861679.1 12 - 13 2346 81
XM_017006191.1 XP_016861680.1 11 - 12 2071 81
XM_017006192.1 XP_016861681.1 12 - 13 1755 81
XM_017006193.1 XP_016861682.1 12 - 13 1752 81
XM_017006194.1 XP_016861683.1 11 - 12 1647 81
XM_017006195.1 XP_016861684.1 6 - 7 1616 81
GenBank mRNA AB371588.1 - 12 - 13 1707 81
AB593126.1 - 11 - 12 1613 81
AK296903.1 - 8 - 9 1193 81
AK298986.1 - 9 - 10 1285 81
AK300437.1 - 12 - 13 1708 81
BC065301.1 - 11 - 12 1510 81
D25542.1 - 12 - 13 1717 81
X75304.1 - 12 - 13 1707 81

View Full Product Details